Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121349.4 | NP_196850.1 | 2 - 3 | 394 | 133 | |
GenBank mRNA | AY054652.1 | - | 2 - 3 | 288 | 133 | |
AY086149.1 | - | 2 - 3 | 243 | 133 | ||
AY128736.1 | - | 1 - 2 | 66 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |