Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BET9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203379.1 | NP_001190308.1 | 6 - 7 | 2039 | 111 | |
NM_001343329.1 | NP_001332743.1 | 7 - 8 | 1817 | 111 | ||
NM_121431.3 | NP_568297.1 | 6 - 7 | 2041 | 111 | ||
GenBank mRNA | AY059785.1 | - | 6 - 7 | 1885 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |