Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343418.1 | NP_001332582.1 | 3 - 4 | 915 | 112 | |
NM_001343419.1 | NP_001332581.1 | 2 - 3 | 994 | 112 | ||
NM_121576.2 | NP_568318.1 | 3 - 4 | 942 | 112 | ||
GenBank mRNA | AY084802.1 | - | 3 - 4 | 637 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |