Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121605.3 | NP_197104.1 | 1 - 2 | 453 | 122 | |
GenBank mRNA | AY074263.1 | - | 1 - 2 | 242 | 122 | |
BT002366.1 | - | 1 - 2 | 123 | 122 | ||
FJ708777.1 | - | 1 - 2 | 123 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |