Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121636.3 | NP_197135.1 | 2 - 3 | 255 | 94 | |
GenBank mRNA | AK118002.1 | - | 2 - 3 | 253 | 94 | |
BT005312.1 | - | 2 - 3 | 123 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |