Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFB1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343479.1 | NP_001331669.1 | 5 - 6 | 705 | 137 | |
NM_121712.4 | NP_197208.1 | 5 - 6 | 705 | 137 | ||
GenBank mRNA | AY086691.1 | - | 5 - 6 | 640 | 137 | |
AY093231.1 | - | 5 - 6 | 550 | 137 | ||
BT003403.1 | - | 5 - 6 | 390 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |