Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ER-ANT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121746.4 | NP_568345.1 | 3 - 4 | 837 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |