Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343592.1 | NP_001332355.1 | 1 - 2 | 238 | 80 | |
NM_121923.4 | NP_568370.1 | 1 - 2 | 238 | 80 | ||
GenBank mRNA | AK226388.1 | - | 1 - 2 | 212 | 80 | |
AY050426.1 | - | 1 - 2 | 175 | 80 | ||
AY120692.1 | - | 1 - 2 | 111 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |