Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336351.1 | NP_001318329.1 | 9 - 10 | 1668 | 66 | |
NM_001336352.1 | NP_001323600.1 | 7 - 8 | 955 | 66 | ||
NM_001336353.1 | NP_001323601.1 | 2 - 3 | 181 | 66 | ||
NM_001336354.1 | NP_001323599.1 | 5 - 6 | 746 | 66 | ||
NM_128748.3 | NP_180749.2 | 7 - 8 | 955 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |