Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128837.1 | NP_180836.1 | 4 - 5 | 1161 | 143 | |
NM_179867.1 | NP_850198.1 | 3 - 4 | 1540 | 143 | ||
NM_201856.4 | NP_973585.1 | 4 - 5 | 1261 | 143 | ||
GenBank mRNA | AF492665.1 | - | 4 - 5 | 1161 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |