Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336416.1 | NP_001318339.1 | 5 - 6 | 1242 | 87 | |
NM_001336417.1 | NP_001325104.1 | 4 - 5 | 737 | 87 | ||
NM_128853.2 | NP_565758.1 | 5 - 6 | 1230 | 87 | ||
GenBank mRNA | AF138744.1 | - | 5 - 6 | 692 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |