Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128874.4 | NP_180873.1 | 13 - 14 | 1698 | 76 | |
GenBank mRNA | AB008854.1 | - | 13 - 14 | 1389 | 76 | |
AF327529.1 | - | 13 - 14 | 1389 | 76 | ||
AF349530.1 | - | 13 - 14 | 1281 | 76 | ||
AY052702.1 | - | 13 - 14 | 1409 | 76 | ||
AY063720.1 | - | 13 - 14 | 1281 | 76 | ||
AY087543.1 | - | 13 - 14 | 1357 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |