Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128999.3 | NP_180994.1 | 1 - 2 | 148 | 71 | |
GenBank mRNA | AK119138.1 | - | 1 - 2 | 113 | 71 | |
AY085812.1 | - | 1 - 2 | 148 | 71 | ||
BT005276.1 | - | 1 - 2 | 54 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |