Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104038.4 | NP_564593.1 | 7 - 8 | 1307 | 94 | |
NM_202273.3 | NP_974002.1 | 7 - 8 | 1069 | 94 | ||
GenBank mRNA | AB119061.1 | - | 7 - 8 | 981 | 94 | |
AY150395.1 | - | 6 - 7 | 840 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |