Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B ALPHA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198270.1 | NP_001185199.1 | 14 - 15 | 1658 | 60 | |
NM_001333494.1 | NP_001322512.1 | 12 - 13 | 3557 | 60 | ||
NM_001333495.1 | NP_001322511.1 | 14 - 15 | 1679 | 60 | ||
NM_104047.4 | NP_564595.1 | 13 - 14 | 3286 | 60 | ||
NM_202274.2 | NP_974003.1 | 12 - 13 | 1580 | 60 | ||
GenBank mRNA | AK230310.1 | - | 13 - 14 | 3066 | 60 | |
AY120756.1 | - | 12 - 13 | 1581 | 60 | ||
BT000109.1 | - | 12 - 13 | 1339 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |