Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQD27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104077.2 | NP_175608.1 | 1 - 2 | 526 | 100 | |
GenBank mRNA | DQ056492.1 | - | 1 - 2 | 345 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |