Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYP73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203224.2 | NP_001190153.1 | 7 - 8 | 810 | 87 | |
NM_001340102.1 | NP_001326528.1 | 6 - 7 | 861 | 87 | ||
NM_001340103.1 | NP_001326529.1 | 7 - 8 | 779 | 87 | ||
NM_116010.1 | NP_567114.1 | 7 - 8 | 766 | 87 | ||
GenBank mRNA | AF355759.1 | - | 7 - 8 | 766 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |