Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116018.4 | NP_191712.1 | 3 - 4 | 802 | 65 | |
NM_202745.4 | NP_974474.1 | 4 - 5 | 1144 | 65 | ||
GenBank mRNA | AY087528.1 | - | 3 - 4 | 802 | 65 | |
BT025255.1 | - | 3 - 4 | 759 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |