Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340110.1 | NP_001327121.1 | 8 - 9 | 1213 | 88 | |
NM_116022.5 | NP_191716.2 | 8 - 9 | 1290 | 88 | ||
GenBank mRNA | AK226722.1 | - | 8 - 9 | 1153 | 88 | |
BT002756.1 | - | 8 - 9 | 994 | 88 | ||
EU249332.1 | - | 8 - 9 | 994 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |