Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035837.1 | NP_001030914.1 | 6 - 7 | 1174 | 118 | |
NM_001340123.1 | NP_001327722.1 | 6 - 7 | 1196 | 118 | ||
NM_116036.4 | NP_567116.1 | 6 - 7 | 1196 | 118 | ||
NM_202746.2 | NP_974475.1 | 6 - 7 | 1182 | 118 | ||
GenBank mRNA | AY039613.1 | - | 6 - 7 | 1182 | 118 | |
BT000508.1 | - | 6 - 7 | 1003 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |