Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RS31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116051.4 | NP_567120.1 | 4 - 5 | 597 | 71 | |
GenBank mRNA | AF439831.1 | - | 4 - 5 | 536 | 71 | |
AY125565.1 | - | 3 - 4 | 452 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |