Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATERDJ3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116125.3 | NP_191819.1 | 4 - 5 | 767 | 109 | |
GenBank mRNA | AY087640.1 | - | 4 - 5 | 628 | 109 | |
AY094429.1 | - | 4 - 5 | 597 | 109 | ||
AY125534.1 | - | 4 - 5 | 464 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |