Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116156.5 | NP_191850.2 | 4 - 5 | 453 | 111 | |
GenBank mRNA | AB109893.1 | - | 4 - 5 | 359 | 111 | |
AY090253.1 | - | 4 - 5 | 400 | 111 | ||
AY125535.1 | - | 4 - 5 | 359 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |