Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125427.1 | NP_001118899.1 | 4 - 5 | 356 | 86 | |
NM_001340197.1 | NP_001326587.1 | 5 - 6 | 395 | 86 | ||
NM_001340198.1 | NP_001326588.1 | 4 - 5 | 395 | 86 | ||
NM_116193.5 | NP_191887.2 | 5 - 6 | 436 | 86 | ||
GenBank mRNA | AY080682.1 | - | 5 - 6 | 430 | 86 | |
AY117234.1 | - | 3 - 4 | 219 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |