Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340236.1 | NP_001328541.1 | 7 - 8 | 1202 | 77 | |
NM_116240.5 | NP_567155.1 | 7 - 8 | 1353 | 77 | ||
GenBank mRNA | AF190794.1 | - | 7 - 8 | 1173 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |