Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203716.1 | NP_001190645.1 | 16 - 17 | 3367 | 89 | |
NM_116288.7 | NP_191972.6 | 16 - 17 | 3400 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |