Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116297.4 | NP_191981.1 | 12 - 13 | 1821 | 63 | |
GenBank mRNA | AK226199.1 | - | 12 - 13 | 1640 | 63 | |
AY062099.1 | - | 12 - 13 | 1633 | 63 | ||
AY124881.1 | - | 12 - 13 | 1344 | 63 | ||
Y07822.1 | - | 1542 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |