Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116297.4 | NP_191981.1 | 7 - 8 | 1347 | 135 | |
GenBank mRNA | AK226199.1 | - | 7 - 8 | 1166 | 135 | |
AY062099.1 | - | 7 - 8 | 1159 | 135 | ||
AY124881.1 | - | 7 - 8 | 870 | 135 | ||
Y07822.1 | - | 1068 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |