Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340295.1 | NP_001319839.1 | 7 - 8 | 2025 | 68 | |
NM_116329.3 | NP_192010.2 | 6 - 7 | 2171 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |