Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIPK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340304.1 | NP_001329151.1 | 6 - 7 | 993 | 68 | |
NM_001340305.1 | NP_001329152.1 | 7 - 8 | 877 | 68 | ||
NM_116349.1 | NP_192028.1 | 6 - 7 | 721 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |