Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340355.1 | NP_001328437.1 | 15 - 16 | 2558 | 62 | |
NM_116433.3 | NP_567221.1 | 16 - 17 | 2496 | 62 | ||
GenBank mRNA | AF100163.1 | - | 16 - 17 | 2450 | 62 | |
AY057477.1 | - | 16 - 17 | 2476 | 62 | ||
AY090293.1 | - | 16 - 17 | 2383 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |