Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMP22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340483.1 | NP_001319865.1 | 2 - 3 | 877 | 72 | |
NM_116685.5 | NP_192356.1 | 1 - 2 | 139 | 72 | ||
GenBank mRNA | AJ006053.1 | - | 1 - 2 | 88 | 72 | |
AK227405.1 | - | 1 - 2 | 108 | 72 | ||
BT003055.1 | - | 1 - 2 | 85 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |