Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203746.1 | NP_001190675.1 | 4 - 5 | 1073 | 94 | |
NM_116723.5 | NP_567272.4 | 4 - 5 | 854 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |