Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203747.1 | NP_001190676.1 | 6 - 7 | 1091 | 115 | |
NM_001340519.1 | NP_001329057.1 | 6 - 7 | 1137 | 115 | ||
NM_116730.5 | NP_192401.5 | 6 - 7 | 804 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |