Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340521.1 | NP_001328262.1 | 10 - 11 | 1001 | 80 | |
NM_116734.3 | NP_567276.1 | 11 - 12 | 1158 | 80 | ||
GenBank mRNA | AY045901.1 | - | 11 - 12 | 1086 | 80 | |
AY113948.1 | - | 11 - 12 | 1065 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |