Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340548.1 | NP_001328043.1 | 7 - 8 | 751 | 125 | |
NM_001340549.1 | NP_001328044.1 | 7 - 8 | 782 | 125 | ||
NM_116772.4 | NP_567292.1 | 7 - 8 | 828 | 125 | ||
GenBank mRNA | AY084733.1 | - | 7 - 8 | 698 | 125 | |
BT024733.1 | - | 6 - 7 | 639 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |