Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TL29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340606.1 | NP_001329837.1 | 5 - 6 | 848 | 104 | |
NM_001340607.1 | NP_001329836.1 | 6 - 7 | 795 | 104 | ||
NM_116970.3 | NP_192640.1 | 6 - 7 | 685 | 104 | ||
GenBank mRNA | AF370534.1 | - | 6 - 7 | 615 | 104 | |
AY072503.1 | - | 6 - 7 | 561 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |