Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116971.6 | NP_192641.2 | 13 - 14 | 1536 | 100 | |
GenBank mRNA | AK227049.1 | - | 13 - 14 | 1373 | 100 | |
AY091058.1 | - | 13 - 14 | 1369 | 100 | ||
AY133739.1 | - | 13 - 14 | 1346 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |