Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116971.6 | NP_192641.2 | 1 - 2 | 296 | 64 | |
GenBank mRNA | AK227049.1 | - | 1 - 2 | 133 | 64 | |
AY091058.1 | - | 1 - 2 | 129 | 64 | ||
AY133739.1 | - | 1 - 2 | 106 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |