Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ISA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116971.6 | NP_192641.2 | 3 - 4 | 402 | 126 | |
GenBank mRNA | AK227049.1 | - | 3 - 4 | 239 | 126 | |
AY091058.1 | - | 3 - 4 | 235 | 126 | ||
AY133739.1 | - | 3 - 4 | 212 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |