Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH7B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104287.5 | NP_175812.1 | 10 - 11 | 1152 | 74 | |
NM_179476.3 | NP_849807.1 | 9 - 10 | 1038 | 74 | ||
GenBank mRNA | AF378873.1 | - | 10 - 11 | 1010 | 74 | |
AJ584645.1 | - | 9 - 10 | 920 | 74 | ||
AK230363.1 | - | 2817 | 74 | |||
AY048242.1 | - | 9 - 10 | 1035 | 74 | ||
AY091032.1 | - | 9 - 10 | 1037 | 74 | ||
AY102145.1 | - | 9 - 10 | 918 | 74 | ||
AY117345.1 | - | 9 - 10 | 918 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |