Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104450.4 | NP_175970.1 | 10 - 11 | 1740 | 80 | |
GenBank mRNA | AK227977.1 | - | 10 - 11 | 1762 | 80 | |
BT004640.1 | - | 10 - 11 | 1669 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |