Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198331.1 | NP_001185260.1 | 1 - 2 | 450 | 83 | |
NM_104600.3 | NP_176115.2 | 1 - 2 | 498 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |