Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104735.4 | NP_176251.1 | 7 - 8 | 1199 | 150 | |
GenBank mRNA | BT000878.1 | - | 7 - 8 | 1000 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |