Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124046.2 | NP_001117518.1 | 4 - 5 | 849 | 150 | |
NM_104743.3 | NP_176259.2 | 4 - 5 | 841 | 150 | ||
GenBank mRNA | AK227267.1 | - | 4 - 5 | 772 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |