Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333920.1 | NP_001322222.1 | 7 - 8 | 671 | 132 | |
NM_001333921.1 | NP_001322221.1 | 1 - 2 | 346 | 132 | ||
NM_104767.4 | NP_176283.1 | 9 - 10 | 793 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |