Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG8C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334017.1 | NP_001319291.1 | 3 - 4 | 431 | 69 | |
NM_104884.5 | NP_176395.1 | 3 - 4 | 431 | 69 | ||
GenBank mRNA | AB073177.1 | - | 3 - 4 | 194 | 69 | |
AK118776.1 | - | 3 - 4 | 290 | 69 | ||
AY085871.1 | - | 3 - 4 | 259 | 69 | ||
BT005441.1 | - | 2 - 3 | 91 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |