Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMO GS-OX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334036.1 | NP_001320894.1 | 3 - 4 | 1008 | 94 | |
NM_001334037.1 | NP_001320895.1 | 3 - 4 | 892 | 94 | ||
NM_104933.3 | NP_564796.1 | 3 - 4 | 997 | 94 | ||
GenBank mRNA | AF370313.1 | - | 3 - 4 | 847 | 94 | |
AY063099.1 | - | 3 - 4 | 772 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |