Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104974.4 | NP_564804.1 | 2 - 3 | 851 | 88 | |
GenBank mRNA | AF348575.1 | - | 2 - 3 | 653 | 88 | |
AY054691.1 | - | 2 - 3 | 691 | 88 | ||
BT008906.1 | - | 2 - 3 | 653 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |