Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNAT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104977.3 | NP_564805.1 | 3 - 4 | 896 | 113 | |
GenBank mRNA | AF308451.1 | - | 3 - 4 | 671 | 113 | |
BT026059.1 | - | 3 - 4 | 595 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |